2-Fluorobenzoic acid, 99%
2-Fluorobenzoic acid, 99%
2-Fluorobenzoic acid, 99%
2-Fluorobenzoic acid, 99%
Thermo Scientific Chemicals
2-Fluorobenzoic acid, 99%
CAS: 445-29-4 | C7H5FO2 | 140.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number FSA119360250
Price (MYR)
293.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
293.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Fluorobenzoic acid
Name Note99%
CAS445-29-4
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning