Fluorobenzene, 99%
Fluorobenzene, 99%
Fluorobenzene, 99%
Thermo Scientific Chemicals
Fluorobenzene, 99%
CAS: 462-06-6 | C6H5F | 96.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA119341000
Price (MYR)
252.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
252.00
BOT
Add to cart
Specifications
Chemical Name or MaterialFluorobenzene
CAS462-06-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.