2-Fluorobenzaldehyde, 97%
2-Fluorobenzaldehyde, 97%
2-Fluorobenzaldehyde, 97%
2-Fluorobenzaldehyde, 97%
Thermo Scientific Chemicals
2-Fluorobenzaldehyde, 97%
CAS: 446-52-6 | C7H5FO | 124.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
Packaging:
Glass bottle
Catalog number FSA119321000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Fluorobenzaldehyde
Name Note97%
CAS446-52-6
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning