Assistent™ Borosilicate Tall Form Beakers
Assistent™ Borosilicate Tall Form Beakers
Actual product may vary
Thermo Scientific Chemicals
Assistent™ Borosilicate Tall Form Beakers
Can be used to make solutions or hold, contain, collect and measure chemicals. Karl Hecht™ Assistent™ Borosilicate Tall Form Beakers are manufactured from durable borosilicate 3.3 glass.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FSA1192910001000 mL
ACR1192910001000 mL
Catalog number FSA119291000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1000 mL
  • Acc. to DIN 12331
  • With pour spout
  • With graduation marks
  • Corrosion-free and resistant to thermal shock
Specifications
Diameter (Metric) Outer95 mm
Capacity (Metric)1000 mL
MaterialBorosilicate Glass
Height (Metric)180 mm
Quantity1 Ea.
TypeBeaker
GraduatedYes
Certifications/ComplianceDIN 12331
StyleTall Form
Unit SizeBOT