3-Fluoroaniline, 98%
3-Fluoroaniline, 98%
3-Fluoroaniline, 98%
3-Fluoroaniline, 98%
Thermo Scientific Chemicals
3-Fluoroaniline, 98%
CAS: 372-19-0 | C6H6FN | 111.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Packaging:
Glass bottle
Catalog number FSA119280250
Price (MYR)
519.00
BOT
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
519.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Fluoroaniline
Name Note98%
CAS372-19-0
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning