2-Fluoroaniline, 99+%
2-Fluoroaniline, 99+%
2-Fluoroaniline, 99+%
2-Fluoroaniline, 99+%
Thermo Scientific Chemicals
2-Fluoroaniline, 99+%
CAS: 348-54-9 | C6H6FN | 111.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Packaging:
Glass bottle
Catalog number FSA119270250
Price (MYR)
288.00
BOT
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
288.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Fluoroaniline
CAS348-54-9
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0