4'-Fluoroacetophenone, 99%
4'-Fluoroacetophenone, 99%
4'-Fluoroacetophenone, 99%
4'-Fluoroacetophenone, 99%
Thermo Scientific Chemicals
4'-Fluoroacetophenone, 99%
CAS: 403-42-9 | C8H7FO | 138.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number FSA119261000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4'-Fluoroacetophenone
Name Note99%
CAS403-42-9
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning