Fluorescein, pure
Fluorescein, pure
Fluorescein, pure
Thermo Scientific Chemicals
Fluorescein, pure
CAS: 2321-07-5 | C20H12O5 | 332.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA119241000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialFluorescein
Melting Point314.0°C to 316.0°C
CAS2321-07-5
Health Hazard 1Warning
Health Hazard 2GHS H Statement: