Flavone, 99+%
Flavone, 99+%
Flavone, 99+%
Thermo Scientific Chemicals
Flavone, 99+%
CAS: 525-82-6 | C15H10O2 | 222.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA119160250
Price (MYR)
2,058.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,058.00
EA
Add to cart
Specifications
Chemical Name or MaterialFlavone
CAS525-82-6
Melting Point95.0°C to 98.0°C
ColorWhite
EINECS Number208-383-8