Ferrocene, 98%
Ferrocene, 98%
Ferrocene, 98%
Thermo Scientific Chemicals
Ferrocene, 98%
CAS: 102-54-5 | C10H10Fe | 186.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA119145000
Price (MYR)
765.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
765.00
BOT
Add to cart
Specifications
Chemical Name or MaterialFerrocene
CAS102-54-5
Health Hazard 1GHS Signal Word: Warning
Melting Point173.0°C to 176.0°C
ColorBrown-Orange