Eugenol, 99%
Eugenol, 99%
Eugenol, 99%
Thermo Scientific Chemicals
Eugenol, 99%
CAS: 97-53-0 | C10H12O2 | 164.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
Glass bottle
Catalog number FSA119115000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialEugenol
Name Note99%
CAS97-53-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement