Ethyl vinyl ether, 99%, stabilized
Ethyl vinyl ether, 99%, stabilized
Ethyl vinyl ether, 99%, stabilized
Ethyl vinyl ether, 99%, stabilized
Thermo Scientific Chemicals
Ethyl vinyl ether, 99%, stabilized
CAS: 109-92-2 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
Packaging:
Glass bottle
Catalog number FSA119082500
Price (MYR)
363.00
EA
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
363.00
EA
Add to cart
Specifications
Chemical Name or MaterialEthyl vinyl ether
Name NoteStabilized
CAS109-92-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapor.
Harmful to aquatic life with long lasting effects.