Ethyltriphenylphosphonium bromide, 98%
Ethyltriphenylphosphonium bromide, 98%
Ethyltriphenylphosphonium bromide, 98%
Ethyltriphenylphosphonium bromide, 98%
Thermo Scientific Chemicals
Ethyltriphenylphosphonium bromide, 98%
CAS: 1530-32-1 | C20H20BrP | 371.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA119041000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialEthyltriphenylphosphonium bromide
Name Note99+%
CAS1530-32-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
View more
for C-C Bond Formation (Olefination)
RUO – Research Use Only