Ethyl pyruvate, 98%
Ethyl pyruvate, 98%
Ethyl pyruvate, 98%
Thermo Scientific Chemicals
Ethyl pyruvate, 98%
CAS: 617-35-6 | C5H8O3 | 116.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA118941000
Price (MYR)
430.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
430.00
BOT
Add to cart
Specifications
Chemical Name or MaterialEthyl pyruvate
Name Note98%
CAS617-35-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Flammable liquid and vapor.