Ethylmercurithiosalicylic acid, sodium salt, 97.0-101.0%
Ethylmercurithiosalicylic acid, sodium salt, 97.0-101.0%
Ethylmercurithiosalicylic acid, sodium salt, 97.0-101.0%
Ethylmercurithiosalicylic acid, sodium salt, 97.0-101.0%
Thermo Scientific Chemicals
Ethylmercurithiosalicylic acid, sodium salt, 97.0-101.0%
Ethylmercurithiosalicylic acid, CAS # 54-64-8, is an oxidizing reagent that can inhibit various sulfhydryl-dependent enzymes and proteins. It is also known as thimerosal or 2-(ethylmercuriomercapto)benzoic acid sodium salt. | CAS: 54-64-8 | C9H9HgNaO2S | 404.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA118685000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialEthylmercurithiosalicylic acid, sodium salt
Melting Point232.0°C to 233.0°C
CAS54-64-8
ColorWhite to Cream
Health Hazard 1GHS Signal Word: Danger
View more
General Description
  • Ethylmercurithiosalicylic acid is an oxidizing reagent that inhibits sulfhydryl-containing active sites of several enzymes
  • It is also known as thimerosal or 2-(ethylmercuriomercapto)benzoic acid sodium salt.
  • It can bind to sulfhydryl compounds, such as glutathione, cysteine, and sulfhydryl groups of proteins
  • Additionally, it can activate inositol triphosphate calcium channels on membranes of the endoplasmic reticulum, resulting in the regulation of cellular functions
Applications
  • Ethylmercurithiosalicylic acid has been widely investigated as a preservative in medicinal laboratory research, including vaccine studies
  • It has been shown to exhibit antifungal and antiseptic properties
RUO – Research Use Only