2-Ethyl-1-hexanol, 99%
2-Ethyl-1-hexanol, 99%
2-Ethyl-1-hexanol, 99%
2-Ethyl-1-hexanol, 99%
Thermo Scientific Chemicals
2-Ethyl-1-hexanol, 99%
CAS: 104-76-7 | C8H18O | 130.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
1 L
2.5 L
Packaging:
View all
Metal drum
Catalog number FSA118530250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 L
Packaging:
Metal drum
Request bulk or custom format
Specifications
Chemical Name or Material2-Ethyl-1-hexanol
CAS104-76-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0