2-Ethylhexanoic acid, 99%
2-Ethylhexanoic acid, 99%
2-Ethylhexanoic acid, 99%
2-Ethylhexanoic acid, 99%
Thermo Scientific Chemicals
2-Ethylhexanoic acid, 99%
CAS: 149-57-5 | C8H16O2 | 144.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
2.5 kg
Packaging:
View all
Glass Bottle
Catalog number FSA118525000
Price (MYR)
223.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
223.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Ethylhexanoic acid
CAS149-57-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0