Ethylene glycol dimethyl ether, 99+%, extra pure, stabilized with BHT
Ethylene glycol dimethyl ether, 99+%, extra pure, stabilized with BHT
Ethylene glycol dimethyl ether, 99+%, extra pure, stabilized with BHT
Ethylene glycol dimethyl ether, 99+%, extra pure, stabilized with BHT
Thermo Scientific Chemicals
Ethylene glycol dimethyl ether, 99+%, extra pure, stabilized with BHT
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number FSA118455000
Price (MYR)
658.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
658.00
BOT
Add to cart
Chemical Identifiers
CAS128-37-0
Specifications
Chemical Name or MaterialEthylene glycol dimethyl ether
pH6 to 7.5 (1:9)
Melting Point-69.0°C
Name NoteStabilized with BHT
Health Hazard 1Danger