Ethylenediamine, 99%
Ethylenediamine, 99%
Ethylenediamine, 99%
Ethylenediamine, 99%
Thermo Scientific Chemicals
Ethylenediamine, 99%
CAS: 107-15-3 | C2H8N2 | 60.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA118420025
Price (MYR)
857.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
857.00
BOT
Add to cart
Specifications
Chemical Name or MaterialEthylenediamine
CAS107-15-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Toxic in contact with skin.
May cause an allergic skin reaction.
Harmful if swallowed.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN (