Ethylene carbonate, 99+%
Ethylene carbonate, 99+%
Ethylene carbonate, 99+%
Ethylene carbonate, 99+%
Ethylene carbonate, 99+%
Thermo Scientific Chemicals
Ethylene carbonate, 99+%
CAS: 96-49-1 | C3H4O3 | 88.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number FSA118412500
Price (MYR)
230.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
230.00
BOT
Add to cart
Specifications
Chemical Name or MaterialEthylene carbonate
CAS96-49-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
Health Hazard 3GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
View more
Suitable for battery materials development.
Alkylation reagent
RUO – Research Use Only