Ethyl chloroacetate, 99%
Ethyl chloroacetate, 99%
Ethyl chloroacetate, 99%
Ethyl chloroacetate, 99%
Thermo Scientific Chemicals
Ethyl chloroacetate, 99%
CAS: 105-39-5 | C4H7ClO2 | 122.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number FSA118222500
Price (MYR)
240.00
EA
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
240.00
EA
Add to cart
Specifications
Chemical Name or MaterialEthyl chloroacetate
CAS105-39-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
Very toxic to aquatic life.
Flammable liquid and vapour.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF INHALED: Remove to fresh air and keep at