Ethyl Benzoate, 99+%
Ethyl Benzoate, 99+%
Ethyl Benzoate, 99+%
Ethyl Benzoate, 99+%
Thermo Scientific Chemicals
Ethyl Benzoate, 99+%
CAS: 93-89-0 | C9H10O2 | 150.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number FSA118095000
Price (MYR)
276.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
276.00
BOT
Add to cart
Specifications
Chemical Name or MaterialEthyl benzoate
Name Note99+%
Acidity1mg KOH/g max.
CAS93-89-0
Health Hazard 1Warning