4-Ethylaniline, 99+%
4-Ethylaniline, 99+%
4-Ethylaniline, 99+%
4-Ethylaniline, 99+%
Thermo Scientific Chemicals
4-Ethylaniline, 99+%
CAS: 589-16-2 | C8H11N | 121.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
Glass bottle
Catalog number FSA118061000
Price (MYR)
637.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
637.00
EA
Add to cart
Specifications
Chemical Name or Material4-Ethylaniline
Name Note99+%
CAS589-16-2
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger