Ethyl acetoacetate, 99%, pure
Ethyl acetoacetate, 99%, pure
Ethyl acetoacetate, 99%, pure
Ethyl acetoacetate, 99%, pure
Thermo Scientific Chemicals
Ethyl acetoacetate, 99%, pure
CAS: 141-97-9 | C6H10O3 | 130.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number FSA117970010
Price (MYR)
391.00
BOT
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
391.00
BOT
Add to cart
Specifications
Chemical Name or MaterialEthyl acetoacetate
Name Note99%
CAS141-97-9
Melting Point-45.0°C