1,2-Ethanedithiol, 95%
1,2-Ethanedithiol, 95%
1,2-Ethanedithiol, 95%
1,2-Ethanedithiol, 95%
Thermo Scientific Chemicals
1,2-Ethanedithiol, 95%
CAS: 540-63-6 | C2H6S2 | 94.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA117852500
Price (MYR)
979.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
979.00
BOT
Add to cart
Specifications
Chemical Name or Material1, 2-Ethanedithiol
CAS540-63-6
CAS Min %94.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0