Estrone, 99+%, Thermo Scientific Chemicals
Estrone, 99+%, Thermo Scientific Chemicals
Estrone, 99+%, Thermo Scientific Chemicals
Estrone, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Estrone, 99+%, Thermo Scientific Chemicals
Estrone, 99%, CAS#53-16-7, is one of the major mammalian estrogen, is a steroid and a sex hormone. It is synthetized from cholesterol and secreted mainly from the gonads. It also presents antineoplastic and bone density conservative activities.
Have Questions?
Catalog NumberQuantityPackaging
FSA1178400101 gGlass bottle
Catalog number FSA117840010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialEstrone
CAS53-16-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Harmful if swallowed.
Suspected of causing cancer.
Harmful in contact with skin.
May cause an allergic skin reaction.
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility.
Health Hazard 3GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
View more
General Description
• Estrone is an aromatized C18 steroid considered one of the major mammalian estrogen and a sex hormone. It shows antineoplastic and bone density conservative activities.
• This compound can interact with estrogen receptors in target tissues to produce similar effects to estradiol. Hormone-bound estrogen receptors dimerize, translocate to the nucleus of cells and bind to estrogen response elements (ERE) of genes. Binding to ERE alters the transcription rate of affected genes.
Applications
• Estrone has the ability to increase the hepatic synthesis of sex hormone binding globulin (SHBG), thyroid-binding globulin (TBG), and other serum proteins.
• It can suppress the release of follicle-stimulating hormone and luteinizing hormone.
• This compound can be produced from androstenedione or testosterone via estradiol. In vivo, it can be generated mainly in the ovaries, placenta, and peripheral tissues (especially adipose tissue) through conversion of androstenedione. It may be metabolized to 16-α-hydroxyestrone, which may be reduced to estriol by estradiol dehydrogenase.
• It has been used as medium supplement for hormone-based degranulation studies of natural killer cells.
• It is used as medium component for monitoring fatty acid synthase activity in breast adenocarcinoma cell lines.
RUO – Research Use Only
General References:
  1. Vardanyan, R.; Hruby, V. 28 - Female Sex Hormones. Synthesis of Essential Drugs. 2006, 365-379.
  2. Macek Jilkova, Z.; Decaens, T.; Marlu, A.; Marche, H.; Jouvin-Marche, E.; Marche, P.N. Sex Differences in Spontaneous Degranulation Activity of Intrahepatic Natural Killer Cells during Chronic Hepatitis B: Association with Estradiol Levels. Mediators Inflamm. 2017, 2017, 3214917.