Emodin, 95%, tech.
Emodin, 95%, tech.
Emodin, 95%, tech.
Thermo Scientific Chemicals
Emodin, 95%, tech.
CAS: 518-82-1 | C15H10O5 | 270.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
Packaging:
Glass bottle
Catalog number FSA117751000
Price (MYR)
991.00
BOT
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
991.00
BOT
Add to cart
Specifications
Chemical Name or MaterialEmodin
Name Notetech., 95%
CAS518-82-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: