Dulcitol, 99+%
Dulcitol, 99+%
Dulcitol, 99+%
Thermo Scientific Chemicals
Dulcitol, 99+%
CAS: 608-66-2 | C6H14O6 | 182.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA117705000
Price (MYR)
2,023.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,023.00
EA
Add to cart
Specifications
Chemical Name or MaterialDulcitol
Name Note99+%
CAS608-66-2
Melting Point188°C to 189°C