Dodecylamine, 98%
Dodecylamine, 98%
Dodecylamine, 98%
Thermo Scientific Chemicals
Dodecylamine, 98%
CAS: 124-22-1 | C12H27N | 185.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
Packaging:
Glass bottle
Catalog number FSA117665000
Price (MYR)
435.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
435.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDodecylamine
Name Note98%
CAS124-22-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquat