1-Dodecanethiol, 98%, Thermo Scientific Chemicals
1-Dodecanethiol, 98%, Thermo Scientific Chemicals
1-Dodecanethiol, 98%, Thermo Scientific Chemicals
1-Dodecanethiol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Dodecanethiol, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA117625000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-Dodecanethiol
CAS112-55-0
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0