1,12-Dodecanediol, 98%
1,12-Dodecanediol, 98%
1,12-Dodecanediol, 98%
1,12-Dodecanediol, 98%
Thermo Scientific Chemicals
1,12-Dodecanediol, 98%
CAS: 5675-51-4 | C12H26O2 | 202.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA117610250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1, 12-Dodecanediol
Name Note99%
CAS5675-51-4
CAS Min %98.5
CAS Max %100.0