2,2'-Dipyridyl, 99+%
2,2'-Dipyridyl, 99+%
2,2'-Dipyridyl, 99+%
2,2'-Dipyridyl, 99+%
Thermo Scientific Chemicals
2,2'-Dipyridyl, 99+%
CAS: 366-18-7 | C10H8N2 | 156.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
10 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA117505000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 2'-Dipyridyl
CAS366-18-7
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0
View more
Indicator
RUO – Research Use Only