Diphenylmethane, 99%
Diphenylmethane, 99%
Diphenylmethane, 99%
Diphenylmethane, 99%
Thermo Scientific Chemicals
Diphenylmethane, 99%
CAS: 101-81-5 | C13H12 | 168.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Packaging:
Glass bottle
Catalog number FSA117355000
Price (MYR)
619.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
619.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDiphenylmethane
Name Note99%
CAS101-81-5
Health Hazard 1GHS Signal Word: Warning
Melting Point24.5°C to 25.0°C