View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1,6-Diphenyl-1,3,5-hexatriene
Name Note98%
CAS1720-32-7
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning