1,1-Diphenylethylene, 98%
1,1-Diphenylethylene, 98%
1,1-Diphenylethylene, 98%
1,1-Diphenylethylene, 98%
Thermo Scientific Chemicals
1,1-Diphenylethylene, 98%
CAS: 530-48-3 | C14H12 | 180.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
25 g
Packaging:
Glass bottle
Catalog number FSA117261000
Price (MYR)
2,069.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,069.00
EA
Add to cart
Specifications
Chemical Name or Material1, 1-Diphenylethylene
CAS530-48-3
CAS Min %97.5
CAS Max %100.0
Melting Point6.0°C