2,2-Diphenylethylamine, 96%
2,2-Diphenylethylamine, 96%
2,2-Diphenylethylamine, 96%
2,2-Diphenylethylamine, 96%
2,2-Diphenylethylamine, 96%
CAS: 3963-62-0 | C14H15N | 197.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA117250050
Price (MYR)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 2-Diphenylethylamine
Name Note96%
CAS3963-62-0
CAS Min %95.0
Health Hazard 1GHS Signal Word: Warning