Diphenyl carbonate, 99%
Diphenyl carbonate, 99%
Diphenyl carbonate, 99%
Diphenyl carbonate, 99%
Thermo Scientific Chemicals
Diphenyl carbonate, 99%
CAS: 102-09-0 | C13H10O3 | 214.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA117231000
Price (MYR)
228.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
228.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDiphenyl carbonate
CAS102-09-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid release to the environment.