N,N'-Diphenylbenzidine, 97%
N,N'-Diphenylbenzidine, 97%
N,N'-Diphenylbenzidine, 97%
N,N'-Diphenylbenzidine, 97%
Thermo Scientific Chemicals
N,N'-Diphenylbenzidine, 97%
CAS: 531-91-9 | C24H20N2 | 336.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
10 g
Packaging:
Glass bottle
Catalog number FSA117210500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialN, N'-Diphenylbenzidine
CAS531-91-9
CAS Min %96.0
CAS Max %100.0
Melting Point246°C to 250°C
View more
Indicator
RUO – Research Use Only