2,4-Dinitrofluorobenzene, 98%
2,4-Dinitrofluorobenzene, 98%
2,4-Dinitrofluorobenzene, 98%
2,4-Dinitrofluorobenzene, 98%
Thermo Scientific Chemicals
2,4-Dinitrofluorobenzene, 98%
CAS: 70-34-8 | C6H3FN2O4 | 186.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA117035000
Price (MYR)
4,930.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
4,930.00
EA
Add to cart
Specifications
Chemical Name or Material2, 4-Dinitrofluorobenzene
CAS70-34-8
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0
View more
Alkylation reagent
RUO – Research Use Only