Lysing Matrix A, 2 mL Tubes
Lysing Matrix A, 2 mL Tubes
Actual product may vary
Lysing Matrix A, 2 mL Tubes
Lysing Matrix A, 2 mL Tubes
Lysing Matrix A, 2 mL Tubes
Lysing Matrix A, 2 mL Tubes
For all sample types except soil for the subsequent isolation of genomic DNA. Lysing Matrix A impact-resistant tubes contain garnet matrix and one 1/4 in. ceramic sphere.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
FSA11691005050 x 2 mL Tubes
ACR11691005050 x 2 mL Tubes
Catalog number FSA116910050
Price (MYR)
-
Quantity:
50 x 2 mL Tubes
  • Tube Size: 2mL
  • Sample Type: Animal tissues, plants, bacteria, yeast, fungi, algae, insects, nematodes
  • Molecule to be Enriched: DNA
  • Grinding matrix is chemically inert and will not bind nucleic acids
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Specifications
Bead TypeGarnet Matrix, 1/4 in. Ceramic Sphere
Sample TypeAnimal Tissues, Plants, Insects, Worms, Bacteria, Yeast, Fungi, Algae
DNaseDnase-free
For Use With (Equipment)FastPrep instruments or other bead beaters
For Use With (Application)Nucleic Acid Isolation and Purification, Homogenization
MatrixLysing Matrix A
Quantity50 x 2 mL Tubes
RNaseRnase-free
Unit SizeBOT
Contents & Storage
Store at Room Temperature.