Dimethyl sulfate, 99+%
Dimethyl sulfate, 99+%
Dimethyl sulfate, 99+%
Dimethyl sulfate, 99+%
Thermo Scientific Chemicals
Dimethyl sulfate, 99+%
CAS: 77-78-1 | C2H6O4S | 126.126 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA1168200101 LPlastic bottle
Catalog number FSA116820010
Price (MYR)
644.00
BOT
Add to cart
Quantity:
1 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
644.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDimethyl sulfate
CAS77-78-1
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
May cause cancer.
Suspected of causing genetic defects.
Fatal if inhaled.
Toxic if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Health Hazard 3GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF exposed or concerned: Get medical advice/attention.
Wear eye protection/face protection.
Obtain special instructions before use.
Avoid breathing dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
View more
Alkylation reagent
RUO – Research Use Only