3,5-Dimethylpyrazole, 99%
3,5-Dimethylpyrazole, 99%
3,5-Dimethylpyrazole, 99%
3,5-Dimethylpyrazole, 99%
Thermo Scientific Chemicals
3,5-Dimethylpyrazole, 99%
CAS: 67-51-6 | C5H8N2 | 96.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA116701000
Price (MYR)
460.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
460.00
BOT
Add to cart
Specifications
Chemical Name or Material3, 5-Dimethylpyrazole
CAS67-51-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0