3,4-Dimethylphenol, 99%
3,4-Dimethylphenol, 99%
3,4-Dimethylphenol, 99%
3,4-Dimethylphenol, 99%
Thermo Scientific Chemicals
3,4-Dimethylphenol, 99%
CAS: 95-65-8 | C8H10O | 122.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA116581000
Price (MYR)
272.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
272.00
BOT
Add to cart
Specifications
Chemical Name or Material3, 4-Dimethylphenol
Name Note99%
CAS95-65-8
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger