Dimethyl methylphosphonate, 97%, Thermo Scientific Chemicals
Dimethyl methylphosphonate, 97%, Thermo Scientific Chemicals
Dimethyl methylphosphonate, 97%, Thermo Scientific Chemicals
Dimethyl methylphosphonate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dimethyl methylphosphonate, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA116420250
Price (MYR)
599.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
599.00
EA
Add to cart
Specifications
Chemical Name or MaterialDimethyl methylphosphonate
CAS756-79-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if inhaled.
Causes serious eye irritation.
May cause genetic defects.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately