N,N-Dimethylformamide, 99+%, extra pure
N,N-Dimethylformamide, 99+%, extra pure
N,N-Dimethylformamide, 99+%, extra pure
N,N-Dimethylformamide, 99+%, extra pure
Thermo Scientific Chemicals
N,N-Dimethylformamide, 99+%, extra pure
CAS: 68-12-2 | C3H7NO | 73.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA116220025
Price (MYR)
501.00
BOT
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
501.00
BOT
Add to cart
Specifications
Chemical Name or MaterialN,N-Dimethylformamide
Melting Point-61.0°C
Name Note99%
CAS68-12-2
CAS Min %99.0