unsym-Dimethylethylenediamine, 99%
unsym-Dimethylethylenediamine, 99%
unsym-Dimethylethylenediamine, 99%
unsym-Dimethylethylenediamine, 99%
Thermo Scientific Chemicals
unsym-Dimethylethylenediamine, 99%
CAS: 108-00-9 | C4H12N2 | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA116200250
Price (MYR)
255.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
255.00
BOT
Add to cart
Specifications
Chemical Name or Materialunsym-Dimethylethylenediamine
CAS108-00-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Highly flammable liquid and vapor.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse