5,5-Dimethyl-1,3-cyclohexanedione, 99%
5,5-Dimethyl-1,3-cyclohexanedione, 99%
5,5-Dimethyl-1,3-cyclohexanedione, 99%
5,5-Dimethyl-1,3-cyclohexanedione, 99%
Thermo Scientific Chemicals
5,5-Dimethyl-1,3-cyclohexanedione, 99%
CAS: 126-81-8 | C8H12O2 | 140.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA116155000
Price (MYR)
1,922.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,922.00
BOT
Add to cart
Specifications
Chemical Name or Material5, 5-Dimethyl-1, 3-cyclohexanedione
Name Note99%
CAS126-81-8
CAS Min %98.5
CAS Max %100.0