2,3-Dimethylbutane, 98+%
2,3-Dimethylbutane, 98+%
2,3-Dimethylbutane, 98+%
2,3-Dimethylbutane, 98+%
Thermo Scientific Chemicals
2,3-Dimethylbutane, 98+%
CAS: 79-29-8 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
View all
Glass bottle
Catalog number FSA116101000
Price (MYR)
731.00
EA
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
731.00
EA
Add to cart
Specifications
Chemical Name or Material2, 3-Dimethylbutane
CAS79-29-8
CAS Min %98.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0