3-Dimethylaminopropylamine, 99%
3-Dimethylaminopropylamine, 99%
3-Dimethylaminopropylamine, 99%
3-Dimethylaminopropylamine, 99%
Thermo Scientific Chemicals
3-Dimethylaminopropylamine, 99%
CAS: 109-55-7 | C5H14N2 | 102.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
5 L
Packaging:
View all
Glass bottle
Catalog number FSA115902500
Price (MYR)
223.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
223.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Dimethylaminopropylamine
CAS109-55-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0