3-Dimethylaminophenol, 97%
3-Dimethylaminophenol, 97%
3-Dimethylaminophenol, 97%
3-Dimethylaminophenol, 97%
Thermo Scientific Chemicals
3-Dimethylaminophenol, 97%
CAS: 99-07-0 | C8H11NO | 137.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA115865000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3-Dimethylaminophenol
CAS99-07-0
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0